Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Methyl Carbonate 98.0+%, TCI America™
SDP

Catalog No. C131525ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
C131525ML 25 mL
C13155ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. C131525ML Supplier TCI America Supplier No. C131525ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 35466-83-2
Molecular Formula C5H8O3
Molecular Weight (g/mol) 116.116
MDL Number MFCD00134545
InChI Key YHLVIDQQTOMBGN-UHFFFAOYSA-N
Synonym allyl methyl carbonate, methyl prop-2-en-1-yl carbonate, carbonic acid allyl methyl ester, isobutylene carbonate, methyl allyl carbonate, acmc-1agon, allyl methyl carbonate #, prop-2-enyl methoxyformate, yhlvidqqtombgn-uhfffaoysa
PubChem CID 534788
IUPAC Name methyl prop-2-enyl carbonate
SMILES COC(=O)OCC=C

Specifications

Color Colorless
Boiling Point 131°C
Quantity 25 mL
UN Number 3272
Formula Weight 116.12
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Methyl Carbonate
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.