Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl methyl sulfide, 98%

Catalog No. AAB2251406
Click to view available options
Quantity:
5 g
25 g

Catalog No. AAB2251406

Supplier: Thermo Scientific Chemicals B2251406

Only null left
Add to Cart

CAS: 10152-76-8 | C4H8S | 88.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 10152-76-8
Molecular Formula C4H8S
Molecular Weight (g/mol) 88.17
MDL Number MFCD00008657
InChI Key NVLPQIPTCCLBEU-UHFFFAOYSA-N
Synonym allyl methyl sulfide, 1-propene, 3-methylthio, allyl methyl sulphide, 3-methylthio-1-propene, sulfide, allyl methyl, methyl propenyl sulfide, unii-v7qi1r316c, methyl allyl sulfide, ccris 7066
PubChem CID 66282
IUPAC Name 3-methylsulfanylprop-1-ene
SMILES CSCC=C

Specifications

Density 0.803
Boiling Point 91°C to 93°C
Flash Point 18°C (65°F)
Odor Stench
Refractive Index 1.4714
Quantity 5 g
UN Number UN1993
Formula Weight 88.17
Percent Purity 98%
Chemical Name or Material Allyl methyl sulfide
Hazard Category H225
Hazard Statement GHS H Statement
H225
Highly flammable liquid and vapour.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 233-422-0
TSCA Yes
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.