Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Nonanoate 97.0+%, TCI America™
SDP

Catalog No. A267925ML
Click to view available options
Quantity:
25 mL

Chemical Identifiers

CAS 7493-72-3
Molecular Formula C12H22O2
Molecular Weight (g/mol) 198.306
MDL Number MFCD00038344
InChI Key MFLWLDDOGSNSKO-UHFFFAOYSA-N
Synonym Nonanoic Acid Allyl Ester, Allyl Pelargonate, Pelargonic Acid Allyl Ester
PubChem CID 61410
IUPAC Name prop-2-enyl nonanoate
SMILES CCCCCCCCC(=O)OCC=C

Specifications

Color Colorless
Boiling Point 151°C
Quantity 25 mL
Formula Weight 198.31
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Nonanoate
EINECSNumber (2)-0759
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.