Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl p-Tolyl Ether 95.0+%, TCI America™
SDP

Catalog No. A128725G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A128725G 25 g
A12875G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. A128725G Supplier TCI America Supplier No. A128725G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 23431-48-3
Molecular Formula C10H12O
Molecular Weight (g/mol) 148.205
MDL Number MFCD00026098
InChI Key AROCNZZBLCAOPH-UHFFFAOYSA-N
Synonym 4-Allyloxytoluene
PubChem CID 262070
IUPAC Name 1-methyl-4-prop-2-enoxybenzene
SMILES CC1=CC=C(C=C1)OCC=C

Specifications

Color Yellow
Quantity 25 g
Formula Weight 148.21
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl p-Tolyl Ether
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.