Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Phenoxyacetate 98.0+%, TCI America™
SDP

Catalog No. A2177250G Shop All Tokyo Chemical Industry (TCI) Products
Click to view available options
Quantity:
25 g
250 g

Chemical Identifiers

CAS 7493-74-5
Molecular Formula C11H12O3
Molecular Weight (g/mol) 192.214
MDL Number MFCD00044632
InChI Key VUFZVGQUAVDKMC-UHFFFAOYSA-N
Synonym allyl phenoxyacetate, allyl 2-phenoxyacetate, acetic acid, phenoxy-, 2-propenyl ester, acetate p.a., acetate pa, phenoxyacetic acid allyl ester, 2-propenyl phenoxyacetate, unii-q3p8uaf9we, acetic acid, phenoxy-, allyl ester, acetate p.a
PubChem CID 24117
IUPAC Name prop-2-enyl 2-phenoxyacetate
SMILES C=CCOC(=O)COC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 137°C
Quantity 250 g
UN Number 2810
Formula Weight 192.21
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Phenoxyacetate
EINECSNumber (3)-0633
RTECSNumber AJ2240000
TSCA Yes
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.