Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyl Phenyl Carbonate 97.0+%, TCI America™
SDP

Catalog No. A230325G
Click to view available options
Quantity:
5 g
25 g

Chemical Identifiers

CAS 16308-68-2
Molecular Formula C10H10O3
Molecular Weight (g/mol) 178.19
MDL Number MFCD07784342
InChI Key ORUWSEKEVGOAQR-UHFFFAOYSA-N
Synonym Carbonic Acid Allyl Phenyl Ester
PubChem CID 11412694
IUPAC Name phenyl prop-2-en-1-yl carbonate
SMILES C=CCOC(=O)OC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 78°C
Quantity 25 g
Formula Weight 178.19
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material Allyl Phenyl Carbonate
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.