Learn More
Allyl trifluoroacetate, 97%
Description
Used as nucleophilic ally metal precursor in alkylation reaction that generates homoallylic amines. Also used as chemical reagent and intermediate for pharmaceutical.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Used as nucleophilic ally metal precursor in alkylation reaction that generates homoallylic amines. Also used as chemical reagent and intermediate for pharmaceutical.
Solubility
Not miscible or difficult to mix with water.
Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
Specifications
Specifications
| Density | 1.183 |
| Boiling Point | 73°C to 75°C |
| Flash Point | −1°C (30°F) |
| Refractive Index | 1.336 |
| Quantity | 10 g |
| UN Number | UN2924 |
| Beilstein | 1766312 |
| Sensitivity | Moisture sensitive |
| Solubility Information | Not miscible or difficult to mix with water. |
| Formula Weight | 154.09 |
| Show More |
Safety and Handling
| Hazard Category | H225-H314 |
| Hazard Statement | GHS H Statement H225-H314-H318 Highly flammable liquid and vapor. Causes severe skin burns and eye damage. Causes serious eye damage. |
| Precautionary Statement | P210-P233-P240-P241-P242-P243-P260-P264b-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P370+P378q-P501c |
| DOTInformation | Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, CORROSIVE, N.O.S. |
| EINECSNumber | 206-853-7 |
| TSCA | No |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Your input is important to us. Please complete this form to provide feedback related to the content on this product.