Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylamine, 98+%

Catalog No. AAL07162AC
Click to view available options
Quantity:
25 mL
100 mL
500 mL

CAS: 107-11-9 | C3H7N | 57.10 g/mol

Allylamine is used as monomer in deposition of the allylamine plasma polymer. It was used in generation of a chemical gradient via plasma co-polymerization of octadiene and allylamine polymer on nanoporous alumina. It was used to investigate the surface reactivity of substrates coated with thin layers of plasma polymerized allylamine films. It was used in a direct monocarboxymethylation reaction.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allylamine is used as monomer in deposition of the allylamine plasma polymer. It was used in generation of a chemical gradient via plasma co-polymerization of octadiene and allylamine polymer on nanoporous alumina. It was used to investigate the surface reactivity of substrates coated with thin layers of plasma polymerized allylamine films. It was used in a direct monocarboxymethylation reaction.

Solubility
Fully miscible in water.

Notes
Air Sensitive. Store away from air and oxidizing agents. Store under dry inert gas. Incompatible with air and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-11-9
Molecular Formula C3H7N
Molecular Weight (g/mol) 57.10
MDL Number MFCD00008199
InChI Key VVJKKWFAADXIJK-UHFFFAOYSA-N
Synonym allylamine, 2-propen-1-amine, monoallylamine, 3-aminopropylene, 3-aminopropene, allyl amine, 2-propenamine, 3-amino-1-propene, 2-propenylamine, polyallylamine
PubChem CID 7853
IUPAC Name prop-2-en-1-amine
SMILES NCC=C

Specifications

Melting Point -88°C
Density 0.761
Boiling Point 53°C to 54°C
Flash Point −28°C (−18°F)
Odor Ammonia-like
Refractive Index 1.421
Linear Formula CH2=CHCH2NH2
Quantity 25 mL
UN Number UN2334
Beilstein 635703
Sensitivity Air Sensitive
Merck Index 14,287
Solubility Information Fully miscible in water.
Formula Weight 57.1
Percent Purity ≥98%
Chemical Name or Material Allylamine
Show More Show Less
Hazard Category H225-H301-H310+H330-H314
Hazard Statement GHS H Statement
H225-H301-H311-H331
Highly flammable liquid and vapor.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P260-P262-P264b-P270-P271-P280-P284-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: ALLYLAMINE
EINECSNumber 203-463-9
RTECSNumber BA5425000
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.