Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylamine Hydrochloride 98.0+%, TCI America™
SDP

Catalog No. A0226100G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 g
100 g
500 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A0226100G 100 g
A022625G 25 g
A0226500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A0226100G Supplier TCI America Supplier No. A0226100G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 10017-11-5
Molecular Formula C3H8ClN
Molecular Weight (g/mol) 93.55
MDL Number MFCD00054329
InChI Key MLGWTHRHHANFCC-UHFFFAOYSA-N
Synonym 3-Aminopropene Hydrochloride
PubChem CID 82291
IUPAC Name hydrogen prop-2-en-1-amine chloride
SMILES [H+].[Cl-].NCC=C

Specifications

Melting Point 106°C
Quantity 100 g
UN Number 2811
Formula Weight 93.55
Percent Purity ≥98.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Allylamine Hydrochloride
EINECSNumber (2)-2379
RTECSNumber BA6950000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.