Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Allylmagnesium chloride, 1.7M solution in THF, AcroSeal™
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
| Color | Black |
| CAS Min % | 15.76, 80.04 |
| CAS Max % | 19.96, 84.25 |
| Linear Formula | H2C=CHCH2MgCl |
| Concentration | 1.5 to 1.9M |
| Density | 0.9600g/mL |
| Synonym | allylmagnesium chloride, magnesium, chloro-2-propenyl, magnesium, chloro-2-propen-1-yl, chloro prop-2-en-1-yl magnesium, allylmagnesium chloride solution, 2.0 m in thf, sjpdadpprujp@, allylmagnesiumchloride, allyl magnesium chloride, allyl-magnesium chloride, 2-propenyl magnesium chloride |
| Solubility Information | Solubility in water: vigorous reaction. |
| IUPAC Name | chloro(prop-2-en-1-yl)magnesium |
| PubChem CID | 75804 |
| Show More |
Safety and Handling
| Hazard Category | Danger |
| Hazard Statement | GHS H Statement Causes severe skin burns and eye damage. May cause respiratory irritation. Suspected of causing cancer. Highly flammable liquid and vapor. Reacts violently with water. May form explosive peroxides. In contact with water releases flammable gases which may ignite spontaneously. May cause drowsiness or dizziness. |
| Precautionary Statement | GHS P Statement Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician. |
RUO – Research Use Only
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction