Alfa Aesar™ Allylmagnesium chloride, 1M in MeTHF

Manufacturer: Alfa Aesar™ H51170AP

Quantity 500mL

 View more versions of this product

Catalog No. AAH51170AP

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 2622-05-1
    Molecular Formula C3H5ClMg
    Linear Formula CH2=CHCH2MgCl
    Formula Weight 100.83
    MDL Number MFCD00000473
    UN Number UN3399
    Chemical Name or Material Allylmagnesium chloride

    Specifications

    Formula Weight 100.83
    TSCA TSCA
    Molecular Formula C3H5ClMg
    Linear Formula CH2=CHCH2MgCl
    MDL Number MFCD00000473
    UN Number UN3399
    Concentration 1M in MeTHF
    Quantity 500mL
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H225-H261-H314-H318
    Highly flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    CAS 2622-05-1
    Health Hazard 3 GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 4.3; Packing Group: II; Proper Shipping Name: ORGANOMETALLIC SUBSTANCE, LIQUID, WATER-REACTIVE, FLAMMABLE
    Chemical Name or Material Allylmagnesium chloride

    Description

    Applications
    Allylmagnesium chloride has been used in a study of the nucleophilic ring-opening of 2-methyleneaziridines to imines and subsequent conversion to 5,5 ft.-disubstituted hydantoins. It is also used as a reagent for the introduction of the allyl group.

    Solubility
    It reacts violently with water.

    Notes
    Air & moisture sensitive. Stable under recommended storage conditions.

    GHS H Statement
    H225-H261-H314-H318
    Highly flammable liquid and vapor.
    In contact with water releases flammable gas.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P210-P231+P232-P303+P361+P353-P305+P351+P338-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    Handle under inert gas.
    Protect from moisture.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit