Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylpentafluorobenzene, 98%

Catalog No. AAA1783406
Click to view available options
Quantity:
5 g

Catalog No. AAA1783406

Supplier: Thermo Scientific Chemicals A1783406

CAS: 1736-60-3 | C9H5F5 | 208.13 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1736-60-3
Molecular Formula C9H5F5
Molecular Weight (g/mol) 208.13
MDL Number MFCD00000301
InChI Key YBDBTBVNQQBHGJ-UHFFFAOYSA-N
Synonym allylpentafluorobenzene, benzene, allylpentafluoro, 3-pentafluorophenyl-1-propene, 2,3,4,5,6-pentafluoro allylbenzene, benzene, pentafluoro-2-propenyl, 1,2,3,4,5-pentafluoro-6-prop-2-en-1-yl benzene, allyl pentafluorobenzene, pubchem13366, acmc-1bth5
PubChem CID 74435
IUPAC Name 1,2,3,4,5-pentafluoro-6-prop-2-enylbenzene
SMILES FC1=C(F)C(F)=C(CC=C)C(F)=C1F

Specifications

Density 1.358
Boiling Point 148°C to 149°C
Flash Point 42°C (108°F)
Refractive Index 1.427
Quantity 5 g
UN Number UN1993
Beilstein 1971209
Formula Weight 208.13
Percent Purity 98%
Chemical Name or Material Allylpentafluorobenzene
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 217-083-6
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.