Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allylthiourea, 98%

Catalog No. AC148800500
Click to view available options
Quantity:
25 g
50 g
100 g
500 g

CAS: 109-57-9 | C4H8N2S | 116.18 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 109-57-9
MDL Number MFCD00004940
InChI Key HTKFORQRBXIQHD-UHFFFAOYSA-N
Synonym allylthiourea, n-allylthiourea, thiosinamine, 1-allyl-2-thiourea, rhodalline, thiosinamin, tiosinamine, 1-allylthiourea, aminosin, rhodallin
PubChem CID 1549517
ChEBI CHEBI:74079
IUPAC Name prop-2-enylthiourea
SMILES C=CCNC(=S)N

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 70°C to 78°C
Color White to Beige
Loss on Drying 1.5% max. (50°C, 2 hrs)
Infrared Spectrum Authentic
Assay Percent Range 98%
Molecular Formula C4H8N2S
Linear Formula H2C=CHCH2NHCSNH2
Quantity 50 g
Beilstein 04, 211
Merck Index 15, 9515
Molecular Weight (g/mol) 116.182
Formula Weight 116.18
Percent Purity 97.5 to 102.5%
Physical Form Crystals, Powder, Granules or Chunks
Chemical Name or Material Allylthiourea, 98%
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement:
Toxic if swallowed.
Precautionary Statement GHS P Statement:
Wash face, hands and any exposed skin thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see .? on this label).

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 203-683-5
RTECSNumber YR8050000
TSCA TSCA

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.