Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyltriethoxysilane, 97%

Catalog No. aal0429714
Click to view available options
Quantity:
5 g
25 g

CAS: 2550-04-1 | C9H20O3Si | 204.34 g/mol

Allyltriethoxysilane is used as chemical additives and intermediates. It is used as general catalyst.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyltriethoxysilane is used as chemical additives and intermediates. It is used as general catalyst.

Solubility
Hydrolyzes in water.

Notes
Moisture Sensitive. Store away from oxidizing agents. Incompatible with moisture and oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2550-04-1
MDL Number MFCD00009064

Specifications

Boiling Point 176°C to 177°C
Quantity 25 g
Molecular Formula C3H6Cl4Si
Linear Formula CH2=CHCH2Si(OCH2CH3)3
UN Number UN1993
Beilstein 1772306
Flash Point 21°C (69°F)
Synonym 3-chloropropyltrichlorosilane, trichloro 3-chloropropyl silane, 3-chloropropyl trichlorosilane, chloropropyltrichlorosilane, silane, trichloro 3-chloropropyl, silane, 3-chloropropyltrichloro, trichloro-3-chloro-propyl-silane, silane, trichloro chloropropyl, g-chloropropyltrichlorosilane, gamma-chloropropyl trichlorosilane
Solubility Information Hydrolyzes in water.
InChI Key OOXSLJBUMMHDKW-UHFFFAOYSA-N
SMILES C(C[Si](Cl)(Cl)Cl)CCl
IUPAC Name trichloro(3-chloropropyl)silane
Molecular Weight (g/mol) 211.97
PubChem CID 75693
Formula Weight 204.34
Percent Purity 97%
Refractive Index 1.407
Sensitivity Moisture sensitive
Density 0.953
Chemical Name or Material Allyltriethoxysilane
Show More Show Less
Hazard Category H225
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapor.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P240-P241-P242-P243-P280-P303+P361+P353-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 219-843-2
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.