Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Allyltrimethylsilane, 98+%

Catalog No. p-7022063
Click to view available options
Quantity:
10 g
50 g
250 g

CAS: 762-72-1 | C6H14Si | 114.263 g/mol

Allyltrimethylsilane is involved as a reactant in Hosomi Sakurai reaction for allylation in the presence of Lewis acid. For example, it reacts with cyclohexanone to get 1-allylcyclohexanol. It acts as a nucleophile and is involved in Carbon-Ferrier rearrangement.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Allyltrimethylsilane is involved as a reactant in Hosomi Sakurai reaction for allylation in the presence of Lewis acid. For example, it reacts with cyclohexanone to get 1-allylcyclohexanol. It acts as a nucleophile and is involved in Carbon-Ferrier rearrangement.

Solubility
Miscible with organic solvents. Immiscible with water.

Notes
Store in cool place. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 762-72-1
Molecular Formula C6H14Si
Molecular Weight (g/mol) 114.263
MDL Number MFCD00008635
InChI Key HYWCXWRMUZYRPH-UHFFFAOYSA-N
Synonym allyltrimethylsilane, silane, trimethyl-2-propenyl, trimethylallylsilane, silane, allyltrimethyl, allyl trimethylsilane, unii-8b84c337vf, ccris 2649, allyl trimethyl silane, 3-trimethylsilyl propene, allytrimethylsilane
PubChem CID 69808
IUPAC Name trimethyl(prop-2-enyl)silane
SMILES C[Si](C)(C)CC=C

Specifications

Boiling Point 85°C to 86°C
Quantity 10 g
UN Number UN1993
Beilstein 906755
Flash Point −10°C (14°F)
Solubility Information Miscible with organic solvents. Immiscible with water.
Formula Weight 114.27
Percent Purity ≥98%
Refractive Index 1.408
Density 0.717
Chemical Name or Material Allyltrimethylsilane
Show More Show Less
Hazard Category H225-H315-H319-H335
Hazard Statement GHS H Statement
H225-H315-H319-H335
Highly flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
EINECSNumber 212-104-5
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.