Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Almond Oil, Sweet, NF, Spectrum™ Chemical

Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
Drum
Steel Can
Description
Spectrum™ Chemical Almond Oil, Sweet, NF is a natural, versatile and effective emollient known for its stability and high level of the "skin-nourishing" Linoleic Acid. Spectrum Chemical NF products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.
Specifications
Specifications
CAS Max % | 100% |
Packaging | Drum |
Quantity | 200 L |
Grade | NF |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction