Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
ALMS1 Antibody, Alexa Fluor™ 750, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP306330AF750
Description
ALMS1 Polyclonal specifically detects ALMS1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
ALMS1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
Alstrom syndrome 1, Alstrom syndrome protein 1, DKFZp686A118, DKFZp686D1828, KIAA0328ALSS | |
Produced in rabbits immunized with E. coli-derived Human ALMS1 fragment. | |
0.1 mL | |
Immune System Diseases, Immunology, Neuronal Cell Markers, Neuroscience, Neurotransmission, Vision | |
7840 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 750 | |
50mM Sodium Borate | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction