Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALMS1 Rabbit anti-Human, Polyclonal, Bethyl Laboratories

Catalog No. 501561064
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-1064 10 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-1064 Supplier Bethyl Laboratories, Inc Supplier No. A301816AT
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

This gene encodes a protein containing a large tandem-repeat domain. The mouse ortholog of this gene has been shown to function in ciliogenesis in inner medullary collecting duct cells. Mutations in this gene have been associated with Alstrom syndrome. Alternative splice variants have been described but their full length sequences have not been determined.

Specifications

Antigen ALMS1
Applications Immunoprecipitation
Classification Polyclonal
Concentration 1 mg/ml
Conjugate Unconjugated
Formulation phosphate, tris citrate with 0.09% sodium azide; pH 7-8
Gene ALMS1
Gene Accession No. Q8TCU4
Gene Alias ALMS1; Alstrom syndrome 1; ALSS; KIAA0328; DKFZp686A118; DKFZp686D1828
Gene Symbols ALMS1
Host Species Rabbit
Immunogen Between 3400 and 3450
Purification Method Antigen affinity chromatography
Quantity 10 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7840
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.