Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Aloin

Catalog No. p-7042328
Click to view available options
Quantity:
100 mg
250 mg
1 g

Aloe exudant, used as a stimulant-laxative | CAS: 1415-73-2 | C21H22O9 | 418.40 g/mol

Aloe exudant, used as a stimulant-laxative

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1415-73-2
Molecular Formula C21H22O9
Molecular Weight (g/mol) 418.40
MDL Number MFCD00151160
InChI Key AFHJQYHRLPMKHU-WEZNYRQKSA-N
Synonym barbaloin, aloin, aloin a, aloinum, aloin ban, dsstox_cid_25967, 10s-1,8-dihydroxy-3-hydroxymethyl-10-2s,3r,4r,5s,6r-3,4,5-trihydroxy-6-hydroxymethyl oxan-2-yl-10h-anthracen-9-one, 1s-1,5-anhydro-1-9s-4,5-dihydroxy-2-hydroxymethyl-10-oxo-9,10-dihydroanthracen-9-yl-d-glucitol, 10-1',5'-anhydroglucosyl aloe-emodin-9-anthrone, barbaloin a
PubChem CID 12305761
ChEBI CHEBI:2991
SMILES OC[C@H]1O[C@H]([C@H](O)[C@@H](O)[C@@H]1O)[C@@H]1C2=CC=CC(O)=C2C(=O)C2=C(O)C=C(CO)C=C12

Specifications

Color Yellow to Brown
Quantity 100 mg
Beilstein 6077558
Merck Index 14,9835
IUPAC Name (10R)-1,8-dihydroxy-3-(hydroxymethyl)-10-[(2S,3R,4R,5S,6R)-3,4,5-trihydroxy-6-(hydroxymethyl)oxan-2-yl]-9,10-dihydroanthracen-9-one
Formula Weight 418.4
Physical Form Powder
Chemical Name or Material Aloin
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
EINECSNumber 215-808-0
RTECSNumber LZ6520000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.