Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALOX12/12 Lipoxygenase Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501985719
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-5719 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-5719 Supplier Bioss Supplier No. BS3874R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Oxygenase and 14,15-leukotriene A4 synthase activity.

Specifications

Antigen ALOX12/12 Lipoxygenase
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene ALOX12
Gene Accession No. P18054
Gene Alias 12(S)-lipoxygenase; 12LO; 12-LO; 12LOX; 12-LOX; 12S LOX; 12S-lipoxygenase; 12S-LOX; 9930022G08Rik; ALOX12; Alox12p; arachidonate 12-lipoxygenase; arachidonate 12-lipoxygenase, 12S type; Arachidonate 12-lipoxygenase, 12S-type; Arachidonate 15-lipoxygenase,15S-type; Linoleate 13S-lipoxygenase; Lipoxin synthase 12-LO; LOG12; P-12LO; platelet 12-LOX; platelet-type 12-lipoxygenase; Platelet-type lipoxygenase 12
Gene Symbols ALOX12
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human 12 Lipoxygenase.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 239
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.