Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALOX12B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA554949
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA554949 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA554949 Supplier Invitrogen™ Supplier No. PA554949
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LYKLLIPHTR YTVQINSIGR AVLLNEGGLS AKGMSLGVEG FAGVMVRALS ELTYDSLYLP NDFVERGVQD LPGYYYRDDS LAVWNALEKY VTEI Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 84%.

This gene encodes an enzyme involved in the converstion of arachidonic acid to 12R-hydroxyeicosatetraenoic acid. Mutations in this gene are associated with nonbullous congenital ichthyosiform erythroderma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALOX12B
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene ALOX12B
Gene Accession No. O75342
Gene Alias 12R-lipoxygenase; 12R-LOX; Alox12b; alox12b {ECO:0000312; Aloxe2; arachidonate 12-lipoxygenase, 12R type; arachidonate 12-lipoxygenase, 12R-type; ARCI2; e-LOX 2; e-LOX2; epidermis-type lipoxygenase 12; Epidermis-type lipoxygenase 2; RGD:1305330}
Gene Symbols ALOX12B
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human ALOX12B. Recombinant protein control fragment (Product #RP-93477).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.