Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALOX12B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5116671
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5116671 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5116671 Supplier Invitrogen™ Supplier No. PA5116671
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: A-431, Mouse spleen, Rat testis Immunogen sequence: MATYKVRVAT GTDLLSGTRD SISLTIVGTQ GESHKQLLNH FGRDFATGAV GQYTVQCPQD LGELIIIRLH KERYAFFPKD PWYCNYVQIC APNGRIYHFP AYQWMDGYET LALREATGKT TADDSLPVLL EHRKEEIRAK QDFYHWRVFL PGLPSYVHIP SYRPPVRRHR NPNRPEWNGY IPGFPILINF KATKFLNLNL RYSFLKTASF FVRLGPMALA FKVRGLLDCK HSWKRLKDIR KIFPGKKSVV.

This gene encodes an enzyme involved in the converstion of arachidonic acid to 12R-hydroxyeicosatetraenoic acid. Mutations in this gene are associated with nonbullous congenital ichthyosiform erythroderma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALOX12B
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 2.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene ALOX12B
Gene Accession No. O70582, O75342, Q2KMM4
Gene Alias 12R-lipoxygenase; 12R-LOX; Alox12b; alox12b {ECO:0000312; Aloxe2; arachidonate 12-lipoxygenase, 12R type; arachidonate 12-lipoxygenase, 12R-type; ARCI2; e-LOX 2; e-LOX2; epidermis-type lipoxygenase 12; Epidermis-type lipoxygenase 2; RGD:1305330}
Gene Symbols ALOX12B
Host Species Rabbit
Immunogen Recombinant protein (or fragment)
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11686, 242, 287425
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.