Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALOX12B Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523608
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523608 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523608 Supplier Invitrogen™ Supplier No. PA523608
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This antibody is predicted to react with mouse and rat based on sequence homology.

This gene encodes an enzyme involved in the converstion of arachidonic acid to 12R-hydroxyeicosatetraenoic acid. Mutations in this gene are associated with nonbullous congenital ichthyosiform erythroderma.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALOX12B
Applications Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.7 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ALOX12B
Gene Accession No. O75342
Gene Alias 12R-lipoxygenase; 12R-LOX; Alox12b; alox12b {ECO:0000312; Aloxe2; arachidonate 12-lipoxygenase, 12R type; arachidonate 12-lipoxygenase, 12R-type; ARCI2; e-LOX 2; e-LOX2; epidermis-type lipoxygenase 12; Epidermis-type lipoxygenase 2; RGD:1305330}
Gene Symbols ALOX12B
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 622-652 amino acids from the C-terminal region of human ALOX12B.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 242
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.