Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALOX15 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577049
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577049 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577049 Supplier Invitrogen™ Supplier No. PA577049
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).

ALOX15 converts arachidonic acid to 15S-hydroperoxyeicosatetraenoic acid. The protein also acts on C-12 of arachidonate as well as on linoleic acid.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALOX15
Applications Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.2
Gene Alox15
Gene Accession No. P16050, Q02759
Gene Alias 12/15-lipoxygenase; 12/15-LO; 12/15-LOX; 12-LO; 12-LOX; 15-lipooxygenase-1; 15-LOX; 15-LOX-1; 15LOX-1; 15-LOX2; Alox12; Alox12l; Alox15; Arachidonate 12-lipoxygenase, leukocyte-type; arachidonate 15-lipoxygenase; Arachidonate omega-6 lipoxygenase; L-12LO; LOG15
Gene Symbols Alox15
Host Species Rabbit
Immunogen A synthesized peptide derived from human 15 Lipoxygenase 1, corresponding to a region within C-terminal amino acids.
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 246, 81639
Target Species Human, Rat, Non-human Primate, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.