Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ALOX15B Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA577050
Description
The antibody was affinity-purified from rabbit antiserum by affinity-chromatography using epitope-specific immunogen and the purity is > 95% (by SDS-PAGE).
ALOX15B encodes a member of the lipoxygenase family of structurally related nonheme iron dioxygenases involved in the production of fatty acid hydroperoxides. The encoded protein converts arachidonic acid exclusively to 15S-hydroperoxyeicosatetraenoic acid, while metabolizing linoleic acid less effectively. This gene is located in a cluster of related genes and a pseudogene that spans approximately 100 kilobases on the short arm of chromosome 17.
Specifications
ALOX15B | |
Polyclonal | |
Unconjugated | |
ALOX15B | |
15-lipoxygenase 2; 15-LOX-2; 15-LOX-B; 15S-lipoxygenase; 8-LOX; 8S-lipoxygenase; 8S-LOX; ALOX15B; Alox8; arachidonate 15-lipoxygenase 2; arachidonate 15-lipoxygenase B; Arachidonate 15-lipoxygenase type II; arachidonate 15-lipoxygenase, second type; arachidonate 15-lipoxygenase, type B; arachidonate 8-lipoxygenase; arachidonate 8S-lipoxygenase; arachidonate omega(6) lipoxygenase; linoleate 13-lipoxygenase 15-LOb | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
11688, 247, 266604 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.2 | |
O15296, O35936, Q8K4F2 | |
ALOX15B, Alox8 | |
Recombinant full length Human ALOX15B. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction