Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALOX15B Recombinant Rabbit Monoclonal Antibody (027)
GREENER_CHOICE

Catalog No. PIMA530056
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA530056 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA530056 Supplier Invitrogen™ Supplier No. MA530056
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). Recombinant rabbit monoclonal antibodies are produced using in vitro expression systems. The expression systems are developed by cloning in the specific antibody DNA sequences from immunoreactive rabbits. Then, individual clones are screened to select the best candidates for production. The advantages of using recombinant rabbit monoclonal antibodies include: better specificity and sensitivity, lot-to-lot consistency, animal origin-free formulations, and broader immunoreactivity to diverse targets due to larger rabbit immune repertoire. This antibody has specificity for Human ALOX15B/15 Lipoxygenase 2.

ALOX15B encodes a member of the lipoxygenase family of structurally related nonheme iron dioxygenases involved in the production of fatty acid hydroperoxides. The encoded protein converts arachidonic acid exclusively to 15S-hydroperoxyeicosatetraenoic acid, while metabolizing linoleic acid less effectively. This gene is located in a cluster of related genes and a pseudogene that spans approximately 100 kilobases on the short arm of chromosome 17.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALOX15B
Applications ELISA
Classification Recombinant Monoclonal
Clone 027
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene ALOX15B
Gene Accession No. O15296
Gene Alias 15-lipoxygenase 2; 15-LOX-2; 15-LOX-B; 15S-lipoxygenase; 8-LOX; 8S-lipoxygenase; 8S-LOX; ALOX15B; Alox8; arachidonate 15-lipoxygenase 2; arachidonate 15-lipoxygenase B; Arachidonate 15-lipoxygenase type II; arachidonate 15-lipoxygenase, second type; arachidonate 15-lipoxygenase, type B; arachidonate 8-lipoxygenase; arachidonate 8S-lipoxygenase; arachidonate omega(6) lipoxygenase; linoleate 13-lipoxygenase 15-LOb
Gene Symbols ALOX15B
Host Species Rabbit
Immunogen Recombinant Human ALOX15B/15 Lipoxygenase 2 Protein (Met1-Ile676).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 247
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.