Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

ALOX5 Monoclonal Antibody (OTI2B7), TrueMAB™, OriGene
SDP

Catalog No. 501681361
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-168-1361 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-168-1361 Supplier OriGene Supplier No. TA807020
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The enzyme 5-lipoxygenase (5-LO) plays a key role in regulating the production of leukotrienes (LT ). Overproduction of LT contributes to several diseases, most notably chronic inflammatory diseases, including asthma, fibrosis and atherosclerosis. Recent work has demonstrated that the activity of 5-LO is regulated by PKA phosphorylation of serine-523 in 5-LO. Under normal conditions, this phosphorylation may be important in limiting inflammation. Abnormal signaling through cAMP and PKA, then, could contribute to a variety of diseases, including those characterized by chronic inflammation.

Specifications

Antigen ALOX5
Applications Western Blot
Classification Monoclonal
Clone OTI2B7
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene ALOX5
Gene Accession No. P09917
Gene Alias 5-LO, 5-LOX, 5LPG, LOG5
Gene Symbols ALOX5
Host Species Mouse
Immunogen Human recombit protein fragment corresponding to amino acids 18-364 of human ALOX5 produced in E.coli.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 240
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.