Learn More
ALOX5 Mouse anti-Human, Clone: OTI2C5, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF807011

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
The enzyme 5-lipoxygenase (5-LO) plays a key role in regulating the production of leukotrienes (LT ). Overproduction of LT contributes to several diseases, most notably chronic inflammatory diseases, including asthma, fibrosis and atherosclerosis. Recent work has demonstrated that the activity of 5-LO is regulated by PKA phosphorylation of serine-523 in 5-LO. Under normal conditions, this phosphorylation may be important in limiting inflammation. Abnormal signaling through cAMP and PKA, then, could contribute to a variety of diseases, including those characterized by chronic inflammation.Specifications
ALOX5 | |
Monoclonal | |
Unconjugated | |
ALOX5 | |
5-LO, 5-LOX, 5LPG, LOG5 | |
Mouse | |
Affinity Chromatography | |
RUO | |
240 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Western Blot | |
OTI2C5 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
P09917 | |
ALOX5 | |
Human recombit protein fragment corresponding to amino acids 18-364 of human ALOX5 produced in E.coli. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.