Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ALOX5 Polyclonal Antibody

Goat Polyclonal Antibody
Supplier: Invitrogen™ PA518567
Description
This antibody is predicted to react with canine and mouse based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
This gene encodes a protein which, with Arachidonate 5-lipoxygenase-activating protein, is required for leukotriene synthesis. Leukotrienes are arachidonic acid metabolites which have been implicated in various types of inflammatory responses, including asthma, arthritis and psoriasis. This protein localizes to the plasma membrane. Inhibitors of its function impede translocation of 5-lipoxygenase from the cytoplasm to the cell membrane and inhibit 5-lipoxygenase activation.
Specifications
| ALOX5 | |
| Polyclonal | |
| Unconjugated | |
| ALOX5 | |
| 5 - Lipoxygenase; 5-lipoxygenase; 5LO; 5-LO; 5-LOX; 5LPG; 5LX; AI850497; ALOX5; Arachidonate 5-lipoxygenase; arachidonic 5-lipoxygenase alpha-10 isoform; arachidonic 5-lipoxygenase delta-10-13 isoform; arachidonic 5-lipoxygenase delta-13 isoform; arachidonic 5-lipoxygenase delta-p10 isoform; arachidonic acid 5-lipoxygenase; F730011J02; leukotriene A4 synthase; LOG5; LOW QUALITY PROTEIN: arachidonate 5-lipoxygenase; lox5; LOX-5; LOX5A; polyunsaturated fatty acid 5-lipoxygenase; RP11-67C2.3 | |
| Goat | |
| Ammonium sulfate precipitation | |
| RUO | |
| 240, 25290 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 | |
| P09917, P12527 | |
| ALOX5 | |
| Synthetic peptide sequence (ERNKKKQLPYYYLSPD) corresponding to the C-terminus amino acids of ALOX5 (aa 651-666). | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction