Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALOXE3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA523953
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA523953 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA523953 Supplier Invitrogen™ Supplier No. PA523953
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

ALOXE3 Polyclonal Antibody for Western Blot, IHC (P)

This gene is a member of the lipoxygenase family, which are catabolized by arachidonic acid-derived compounds. The encoded enzyme is a hydroperoxide isomerase that synthesizes a unique type of epoxy alcohol (8R-hydroxy-11R,12R-epoxyeicosa-5Z,9E,14Z-trienoic acid) from 12R-hydroperoxyeicosatetraenoic acid (12R-HPETE). This epoxy alcohol can activate the the nuclear receptor peroxisome proliferator-activated receptor alpha (PPARalpha), which is implicated in epidermal differentiation. Loss of function of the enzyme encoded by this gene results in ichthyosis, implicating the function of this gene in the differentiation of human skin. This gene is part of a cluster of lipoxygenase genes on 17p13.1. Mutations in this gene result in nonbullous congenital ichthyosiform erythroderma (NCIE). Multiple transcript variants encoding different isoforms have been found for this gene.[provided by RefSeq, Sep 2009].
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALOXE3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene ALOXE3
Gene Accession No. Q9BYJ1
Gene Alias ALOXE3; arachidonate lipoxygenase 3; ARCI3; E-LOX; eLOX3; e-LOX-3; eLOX-3; epidermal lipoxygenase; epidermal LOX-3; Epidermis-type lipoxygenase 3; hydroperoxide isomerase ALOXE3; Hydroperoxy icosatetraenoate dehydratase
Gene Symbols ALOXE3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 223-252 amino acids from the central region of human ALOXE3.
Purification Method Protein A, Antigen affinity chromatography
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 59344
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.