Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALP Monoclonal Antibody (7B6)
GREENER_CHOICE

Catalog No. PIMA533328
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA533328 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA533328 Supplier Invitrogen™ Supplier No. MA533328
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

7B6 reacts with CRASH, a 308 AA glycoprotein sharing 32% homology with human asparaginase and identical to Asparaginase-Like Protein (ALP) found in rat sperm. CRASH is found in a variety of tumors but only occasionally in normal tissues. Especially ovarian cancers are highly positive. CRASH is further found in brain tumors, some colonic cancers, some lung cancers, some prostate cancers, uterine cancers, some breast cancers and some thyroid cancers. It was not found in serum. In Western blot 3 bands are demonstrated of 45 kDa, 28 kDa and 15 kDa.

ALP is found in a variety of tumors but only occasionally in normal tissues. Ovarian cancers are highly positive. ALP is further found in brain tumors, some colonic cancers, some lung cancers, some prostate cancers, uterine cancers, some breast cancers and some thyroid cancers. It was not found in serum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALP
Applications ELISA, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone 7B6
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.02% sodium azide
Gene Asrgl1
Gene Accession No. Q7L266
Gene Alias 2410004D18Rik; ALP; ALP1; asparaginase and isoaspartyl peptidase 1; asparaginase like 1; asparaginase-like 1; asparaginase-like 1 protein; asparaginase-like protein 1; Asparaginase-like sperm autoantigen; ASRGL1; AU040643; AW060375; Beta-aspartyl-peptidase; CRASH; FLJ22316; Glial asparaginase; Gliap; Hiob; Hiob2; isoaspartyl dipeptidase; Isoaspartyl peptidase/L-asparaginase; Isoaspartyl peptidase/L-asparaginase alpha chain; Isoaspartyl peptidase/L-asparaginase beta chain; L-asparaginase; L-asparagine amidohydrolase; testis secretory sperm-binding protein Li 242mP; zgc:103568
Gene Symbols Asrgl1
Host Species Mouse
Immunogen CRASH recombinant protein and synthetic peptide.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80150
Target Species Human
Content And Storage 4°C, do not freeze
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.