Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ ALP Monoclonal Antibody (CL1679)
GREENER_CHOICE

Catalog No. PIMA524629
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA524629 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA524629 Supplier Invitrogen™ Supplier No. MA524629
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Immunogen sequence: MDGKDLSAGA VSAVQCIANP IKLARLVMEK TPHCFLTDQG AAQFAAAMGV PEIPGEKLVT ERNKKRLEKE KHEKGAQKTD CQ Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 79%. Binds to an epitope located within the peptide sequence TPHCFLTDQGAAQFA as determined by overlapping synthetic peptides.

ALP is found in a variety of tumors but only occasionally in normal tissues. Ovarian cancers are highly positive. ALP is further found in brain tumors, some colonic cancers, some lung cancers, some prostate cancers, uterine cancers, some breast cancers and some thyroid cancers. It was not found in serum.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ALP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Monoclonal
Clone CL1679
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene Asrgl1
Gene Accession No. Q7L266
Gene Alias 2410004D18Rik; ALP; ALP1; asparaginase and isoaspartyl peptidase 1; asparaginase like 1; asparaginase-like 1; asparaginase-like 1 protein; asparaginase-like protein 1; Asparaginase-like sperm autoantigen; ASRGL1; AU040643; AW060375; Beta-aspartyl-peptidase; CRASH; FLJ22316; Glial asparaginase; Gliap; Hiob; Hiob2; isoaspartyl dipeptidase; Isoaspartyl peptidase/L-asparaginase; Isoaspartyl peptidase/L-asparaginase alpha chain; Isoaspartyl peptidase/L-asparaginase beta chain; L-asparaginase; L-asparagine amidohydrolase; testis secretory sperm-binding protein Li 242mP; zgc:103568
Gene Symbols Asrgl1
Host Species Mouse
Immunogen Recombinant protein fragment of Human ASRGL1 (Product #RP-95864)
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 80150
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.