Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ ASZ1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA535858
Description
Store product as a concentrated solution. Centrifuge briefly prior to opening the vial. Predicted reactivity: Mouse (83%), Rat (84%), Cat (89%), Rabbit (89%), Sheep (86%), Rhesus Monkey (97%), Chimpanzee (99%), Bovine (87%).
Plays a central role during spermatogenesis by repressing transposable elements and preventing their mobilization, which is essential for the germline integrity. Acts via the piRNA metabolic process, which mediates the repression of transposable elements during meiosis by forming complexes composed of piRNAs and Piwi proteins and governs the methylation and subsequent repression of transposons. Its association with pi-bodies suggests a participation in the primary piRNAs metabolic process. Required prior to the pachytene stage to facilitate the production of multiple types of piRNAs, including those associated with repeats involved in the regulation of retrotransposons. May act by mediating protein-protein interactions during germ cell maturation.
Specifications
| ASZ1 | |
| Polyclonal | |
| Unconjugated | |
| Asz1 | |
| 4933400N19Rik; ALP1; ANKL1; ankyrin repeat, SAM and basic leucine zipper domain containing 1; ankyrin repeat, SAM and basic leucine zipper domain-containing protein 1; ankyrin-like 1; ankyrin-like protein 1; Asz1; C7orf7; CT1.19; GASZ; germ cell-specific ankyrin, SAM and basic leucine zipper domain containing protein; germ cell-specific ankyrin, SAM and basic leucine zipper domain-containing protein; Orf3 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 136991, 74068 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.77 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q8VD46, Q8WWH4 | |
| Asz1 | |
| Recombinant protein encompassing a sequence within the center region of human ASZ1. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction