Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-1-Anti-Trypsin Polyclonal Antibody, Rockland™
SDP

Catalog No. 501991678
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
100 μg
25 μL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-199-1678 100 μg
50-199-1679 25 μL
50-199-1677 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. 50-199-1678 Supplier Rockland Immunochemicals Supplier No. 6004011470100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Store vial at -20°C prior to opening. Aliquot contents and freeze at -20°C or below for extended storage. Avoid cycles of freezing and thawing. Centrifuge product if not completely clear after standing at room temperature. This product is stable for several weeks at 4°C as an undiluted liquid. Dilute only prior to immediate use. Assay by immunoelectrophoresis resulted in a single precipitin arc against anti-Rabbit Serum. Analysis by SDS-PAGE was used to determine that this preparation is substantially free of aggregates and shows a banding pattern consistent with purified Rabbit IgG.

Alpha-1-AT is synthesized in the liver and it acts as an inhibitor of proteases such as trypsin, elastase, chymotrypsin, collagenase, leucocytic proteases, plasmin, and thrombin, which may be released during inflammatory reactions in the lung. In the absence of alpha-1-AT, these enzymes are not inhibited and they may digest pulmonary parenchyma. Alpha-1-AT deficiency is associated with chronic obstructive lung disease (emphysema) and less frequently with hepatic cirrhosis in infants and respiratory distress of the newborn. Increase in alpha-1-AT occurs as an acute phase response to tissue necrosis and inflammation. Serum level of alpha-1-AT is elevated in rheumatoid arthritis, bacterial infections, vasculitis, and carcinomatosis.

Specifications

Antigen alpha-1-Anti-Trypsin
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation 0.02M potassium phosphate with 0.15M NaCl and 0.01% sodium azide; pH 7.2
Gene SERPINA1
Gene Accession No. P01009
Gene Alias a-1 AT; A1A; a1-antitrypsin; A1AT; AAT; Aat2; Aat-2; alpha-1 antitrypsin; Alpha-1 protease inhibitor; Alpha-1 protease inhibitor 1; Alpha-1-antiproteinase; Alpha-1-antitrypsin; alpha-1-antitrypsin (protease inhibitor); Alpha-1-antitrypsin 1-1; alpha-1-antitrypsin 2; alpha-1-antitrypsin null; alpha1AT; alpha-1-protease inhibitor; Alpha-1-proteinase inhibitor; Digestive Zymogen; Dom1; MGC23330; MGC9222; PI; PI1; PRO0684; PRO2209; PRO2275; protease inhibitor 1 (anti-elastase), alpha-1-antitrypsin; Protease Serine 1; serine (or cysteine) peptidase inhibitor, clade A, member 1A; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1a; Serine Protease 1; Serine protease inhibitor 1-1; Serine protease inhibitor A1a; serine protease inhibitor alpha 1; Serpin A1; Serpin A1a; serpin family A member 1; serpin peptidase inhibitor clade A member 1; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1; SERPINA1; Serpina1a; Short peptide from AAT; SPAAT; Spi1; Spi1-1; Spi1-3; TRP1; TRY1
Gene Symbols SERPINA1
Host Species Rabbit
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5265
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.