Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha-1-Antitrypsin Mouse anti-Human, Rat, Clone: 1A9G6, Proteintech
SDP

Catalog No. 501736571 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-6571 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-6571 Supplier Proteintech Group Inc Supplier No. 661351IG150UL
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The serum glycoprotein a1-AT, is a typical member of the serpin family (serine protease inhibitors), and, an inhibitor of trypsin. a1- AT is synthesized in the liver thus as a single polypeptide chain of approximately 51 kDa showing a marked degree of genetic polymorphism due to the presence of over 20 different alleles. Individuals homozygous for a1-AT defiiency are predisposed to pulmonary emphysema and hepatic cirrhosis.

Specifications

Antigen Alpha-1-Antitrypsin
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone 1A9G6
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SERPINA1
Gene Accession No. P01009, P17475
Gene Alias A1A, A1AT, AAT, Alpha 1 antiproteinase, Alpha 1 antitrypsin, Alpha 1 protease inhibitor, PI, PI1, PRO2209, PRO2275, Serpin A1, SERPINA1, SPAAT, alpha1-antitrypsin
Gene Symbols SERPINA1
Host Species Mouse
Immunogen Alpha-1-Antitrypsin Fusion Protein Ag9516
Purification Method Protein G
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 24648, 5265
Target Species Human, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.