Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha-1 Antitrypsin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA585339
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA585339 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA585339 Supplier Invitrogen™ Supplier No. PA585339
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Keep as concentrated solution. Predicted reactivity: Rhesus Monkey (90%). Positive Control: A549, HepG2, Human lung cancer, Human cervical carcinoma. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

Alpha-1-AT is synthesized in the liver and it acts as an inhibitor of proteases such as trypsin, elastase, chymotrypsin, collagenase, leucocytic proteases, plasmin, and thrombin, which may be released during inflammatory reactions in the lung. In the absence of alpha-1-AT, these enzymes are not inhibited and they may digest pulmonary parenchyma. Alpha-1-AT deficiency is associated with chronic obstructive lung disease (emphysema) and less frequently with hepatic cirrhosis in infants and respiratory distress of the newborn. Increase in alpha-1-AT occurs as an acute phase response to tissue necrosis and inflammation. Serum level of alpha-1-AT is elevated in rheumatoid arthritis, bacterial infections, vasculitis, and carcinomatosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-1 Antitrypsin
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.81 mg/mL
Conjugate Unconjugated
Formulation 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7
Gene SERPINA1
Gene Accession No. P01009
Gene Alias a-1 AT; A1A; a1-antitrypsin; A1AT; AAT; Aat2; Aat-2; alpha 1 antitrypsin; alpha-1 antitrypsin; alpha-1 protease inhibitor; Alpha-1 protease inhibitor 1; alpha-1-antiproteinase; Alpha-1-antitrypsin; alpha-1-antitrypsin (protease inhibitor); alpha-1-antitrypsin 1-1; alpha-1-antitrypsin 2; alpha-1-antitrypsin null; alpha1AT; alpha-1-protease inhibitor; alpha-1-proteinase inhibitor; Digestive Zymogen; Dom1; MGC23330; MGC9222; PI; PI1; PRO0684; PRO2209; PRO2275; protease inhibitor 1 (anti-elastase), alpha-1-antitrypsin; protease inhibitor 1 (anti-elastase, antitrypsin, alpha-1); Protease Serine 1; serine (or cysteine) peptidase inhibitor, clade A, member 1A; serine (or cysteine) proteinase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1; serine (or cysteine) proteinase inhibitor, clade A (alpha-1antiproteinase, antitrypsin), member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1; serine (or cysteine) proteinase inhibitor, clade A, member 1a; Serine Protease 1; serine protease inhibitor 1-1; Serine protease inhibitor A1a; serine protease inhibitor alpha 1; serine proteinase inhibitor, clade A, member 1; serpin A1; Serpin A1a; serpin family A member 1; serpin peptidase inhibitor clade A member 1; serpin peptidase inhibitor, clade A (alpha-1 antiproteinase, antitrypsin), member 1; SERPINA1; Serpina1a; Short peptide from AAT; SPAAT; Spi1; Spi1-1; Spi1-3; TRP1; TRY1
Gene Symbols SERPINA1
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human alpha 1 Antitrypsin. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5265
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.