Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha-1-Antitrypsin Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
$518.60
Specifications
Antigen | Alpha-1-Antitrypsin |
---|---|
Concentration | 0.48 mg/mL |
Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot |
Classification | Polyclonal |
Conjugate | Unconjugated |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-554-071
![]() ![]() |
Proteintech Group Inc
163821AP150UL |
150 μL |
Each of 1 for $518.60
|
|
|||||
Description
The serum glycoprotein a1-AT, is a typical member of the serpin family (serine protease inhibitors), and, an inhibitor of trypsin. a1- AT is synthesized in the liver thus as a single polypeptide chain of approximately 51 kDa showing a marked degree of genetic polymorphism due to the presence of over 20 different alleles. Individuals homozygous for a1-AT defiiency are predisposed to pulmonary emphysema and hepatic cirrhosis.Specifications
Alpha-1-Antitrypsin | |
Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
Unconjugated | |
Rabbit | |
Human, Mouse, Rat | |
P01009, P07758, P17475 | |
20700, 24648, 5265 | |
Alpha-1-Antitrypsin Fusion Protein Ag9369 | |
Primary | |
-20°C | |
SERPINA1 |
0.48 mg/mL | |
Polyclonal | |
Liquid | |
RUO | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
A1A, A1AT, AAT, Alpha 1 antiproteinase, Alpha 1 antitrypsin, Alpha 1 protease inhibitor, PI, PI1, PRO2209, PRO2275, Serpin A1, SERPINA1, SPAAT, alpha1-antitrypsin | |
SERPINA1, Serpina1a | |
IgG | |
Antigen Affinity Chromatography | |
Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title