Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha 1 Fetoprotein Receptor Antibody (5E1), Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP278787
Description
Alpha 1 Fetoprotein Receptor Monoclonal specifically detects Alpha 1 Fetoprotein Receptor in Human samples. It is validated for Western Blot, Immunohistochemistry.Specifications
Alpha 1 Fetoprotein Receptor | |
Monoclonal | |
Unconjugated | |
PBS, pH 7.4 with 0.09% Sodium Azide | |
Mouse | |
Protein A purified | |
Primary | |
Human | |
Purified |
Western Blot, Immunohistochemistry | |
50 | |
Western Blot, Immunohistochemistry | |
Alpha-1-fetoprotein, Alpha-fetoglobulin, alpha-fetoprotein, FETA, HP, HPAFP | |
Full length native protein (purified) (Human). | |
0.2 mg | |
174 | |
Store at 4°C. Do not freeze. | |
IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction