Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ alpha-1 Microglobulin Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA580336
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA580336 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA580336 Supplier Invitrogen™ Supplier No. PA580336
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

This product is preservative free. It is recommended to add sodium azide to avoid contamination (final concentration 0.05%-0.1%). This antibody has specificity for Human AMBP/Alpha 1 microglobulin.

This gene encodes a complex glycoprotein secreted in plasma. The precursor is proteolytically processed into distinct functioning proteins: alpha-1-microglobulin, which belongs to the superfamily of lipocalin transport proteins and may play a role in the regulation of inflammatory processes, and bikunin, which is a urinary trypsin inhibitor belonging to the superfamily of Kunitz-type protease inhibitors and plays an important role in many physiological and pathological processes. This gene is located on chromosome 9 in a cluster of lipocalin genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-1 Microglobulin
Applications ELISA, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene AMBP
Gene Accession No. P02760
Gene Alias A1M; AI194774; alpha 1 microglobulin/bikunin; alpha-1 microglobulin; Alpha-1 microglobulin/bikunin; Alpha-1 microglycoprotein; Alpha-1-microglobulin; alpha-1-microglobulin/bikunin; alpha1-microglobulin/bikunin; alpha-1-microglobulin/bikunin precursor; alpha-1-microglobulin-bikunin; alpha1-microglobulin-bikunin precursor; Ambp; AMBP protein; ASPI; BI-14; Bikunin; complex-forming glycoprotein heterogeneous in charge; Cumulus extracellular matrix-stabilizing factor; EDC1; ESF; growth-inhibiting protein 19; HCP; HI30; HI-30; IATIL; inter-alpha-trypsin inhibitor (protein HC), light; inter-alpha-trypsin inhibitor light chain; Intin4; ITI; ITIL; ITILC; ITI-LC; polyprotein; protein AMBP; Protein HC; Protein Protein HC; trypstatin; ulinastatin; unnamed protein product; Urinary Trypsin Inhibitor; uristatin; uronic-acid-rich protein; UTI
Gene Symbols AMBP
Host Species Rabbit
Immunogen Recombinant Human AMBP/Alpha 1 microglobulin protein (Met1-Val203).
Purification Method Antigen affinity chromatography, Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 259
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.