Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha 1 microglobulin Rabbit anti-Human, Polyclonal, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6512R

Description
This gene encodes a complex glycoprotein secreted in plasma. The precursor is proteolytically processed into distinct functioning proteins: alpha-1-microglobulin, which belongs to the superfamily of lipocalin transport proteins and may play a role in the regulation of inflammatory processes, and bikunin, which is a urinary trypsin inhibitor belonging to the superfamily of Kunitz-type protease inhibitors and plays an important role in many physiological and pathological processes. This gene is located on chromosome 9 in a cluster of lipocalin genes.Specifications
Alpha 1 microglobulin | |
Polyclonal | |
Unconjugated | |
AMBP | |
A1M; AI194774; alpha 1 microglobulin/bikunin; alpha-1 microglobulin; Alpha-1 microglobulin/bikunin; Alpha-1 microglycoprotein; Alpha-1-microglobulin; alpha-1-microglobulin/bikunin; alpha-1-microglobulin/bikunin precursor; alpha-1-microglobulin/bikunin-like; alpha-1-microglobulin-bikunin; Ambp; AMBP protein; ASPI; BI-14; Bikunin; BOS_9342; Complex-forming glycoprotein heterogeneous in charge; Cumulus extracellular matrix-stabilizing factor; EDC1; ESF; growth-inhibiting protein 19; HCP; HI30; HI-30; IATIL; inter-alpha-trypsin inhibitor (protein HC), light; Inter-alpha-trypsin inhibitor light chain; Intin4; ITI; Itil; ITILC; ITI-LC; LOC100356142; polyprotein; Protein AMBP; Protein HC; Protein Protein HC; Trypstatin; ulinastatin; Urinary Trypsin Inhibitor; uristatin; Uronic-acid-rich protein; UTI | |
Rabbit | |
Protein A | |
RUO | |
259 | |
-20°C | |
Liquid |
Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot | |
1 μg/mL | |
PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH | |
P02760 | |
AMBP | |
KLH conjugated synthetic peptide derived from human Protein AMBP . | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction