Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-1B Adrenergic R/ADRA1B Antibody (471802), Alexa Fluor™ 350, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB4730U
Description
alpha-1B Adrenergic R/ADRA1B Monoclonal antibody specifically detects alpha-1B Adrenergic R/ADRA1B in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
alpha-1B Adrenergic R/ADRA1B | |
Monoclonal | |
Alexa Fluor 350 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
Primary | |
Human | |
Purified |
Flow Cytometry, CyTOF | |
471802 | |
Intracellular Staining by Flow Cytometry, CyTOF-ready | |
ADRA1, adrenergic, alpha-1B-, receptor, alpha-1B adrenergic receptor, Alpha-1B adrenoceptor, Alpha-1B adrenoreceptor, alpha-1B-adrenergic receptor, ALPHA1BAR | |
NS0 mouse myeloma cell line transfected with human ADRA1B, Met1-Phe520, Accession # P35368 | |
0.1 mL | |
GPCR, Neuroscience | |
147 | |
Store at 4C in the dark. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction