Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Human alpha-1B Adrenergic R/ADRA1B Antibody, R&D Systems™

Mouse Monoclonal Antibody
Supplier: R&D Systems MAB4730SP
This item is not returnable.
View return policy
Description
alpha-1B Adrenergic R/ADRA1B Monoclonal specifically detects alpha-1B Adrenergic R/ADRA1B in Human samples. It is validated for Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
alpha-1B Adrenergic R/ADRA1B | |
Monoclonal | |
LYOPH | |
Intracellular Staining by Flow Cytometry 2.5 ug/10^6 cells | |
P35368 | |
ADRA1B | |
NS0 mouse myeloma cell line transfected with human ADRA1B Met1-Phe520 Accession # P35368 | |
25 μg | |
Primary | |
Detects human ADRA1B. Stains human ADRA1B transfectants but not irrelevant transfectants. | |
Human | |
Purified |
Flow Cytometry | |
471802 | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
a-1B AdrenergicR, ADRA1, ADRA1B, adrenergic, alpha-1B-, receptor, alpha-1B adrenergic receptor, alpha1B AdrenergicR, Alpha-1B adrenoceptor, Alpha-1B adrenoreceptor, alpha-1B-adrenergic receptor, ALPHA1BAR | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
147 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction