Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-1b Adrenergic Receptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5114827
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5114827 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5114827 Supplier Thermo Scientific Supplier No. PA5114827
Add to Cart
Add to Cart

alpha-1b Adrenergic Receptor Polyclonal Antibody

alpha-1b Adrenergic Receptor Polyclonal Antibody for Western Blot, ICC/IF

TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-1b Adrenergic Receptor
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene Adra1b
Gene Accession No. P15823, P35368, P97717
Gene Alias [a]1b; ADA1B; ADRA1; ADRA1B; Adrenergic alpha 1B- receptor; adrenergic receptor, alpha 1b; adrenergic, alpha-1B-, receptor; adrenoceptor alpha 1B; alpha 1B-adrenoceptor; alpha 1B-adrenoreceptor; alpha-1B adrenergic receptor; Alpha-1B adrenoceptor; alpha-1B adrenoreceptor; alpha-1B-adrenergic receptor; alpha1B-adrenergic receptor; ALPHA1BAR
Gene Symbols Adra1b
Host Species Rabbit
Immunogen A synthesized peptide derived from human ADRA1B(Accession P35368), corresponding to amino acid residues R403-L453.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11548, 147, 24173
Target Species Human, Mouse, Rat, Monkey
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.