Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha-1D Adrenergic R/ADRA1D Antibody, PE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP297580PE
Description
alpha-1D Adrenergic R/ADRA1D Polyclonal antibody specifically detects alpha-1D Adrenergic R/ADRA1D in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
alpha-1D Adrenergic R/ADRA1D | |
Polyclonal | |
Immunohistochemistry-Paraffin | |
ADRA1, ADRA1ADAR, ADRA1R, adrenergic, alpha -1D-, receptor, adrenergic, alpha-1A-, receptor, adrenergic, alpha-1D-, receptor, ALPHA1, Alpha-1A adrenergic receptor, alpha-1D adrenergic receptor, Alpha-1D adrenoceptor, Alpha-1D adrenoreceptor, alpha-1D-adrenergic receptor, Alpha-adrenergic receptor 1a, dJ779E11.2 | |
Produced in rabbits immunized with E. coli-derived Human alpha-1D Adrenergic R/ADRA1D fragment. | |
0.1 mL | |
Cell Cycle and Replication, GPCR, Neuroscience | |
146 | |
Store at 4C in the dark. | |
IgG |
Immunohistochemistry (Paraffin) | |
PE | |
PBS | |
Rabbit | |
Antigen and protein A Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction