Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha-1d Adrenoceptor Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA533328
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA533328 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA533328 Supplier Thermo Scientific Supplier No. PA533328
Add to Cart
Add to Cart

alpha-1d Adrenoceptor Polyclonal Antibody

alpha-1d Adrenoceptor Polyclonal Antibody for IHC (P)

TRUSTED_SUSTAINABILITY

Specifications

Antigen alpha-1d Adrenoceptor
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide
Gene Adra1d
Gene Accession No. O02666, P25100, Q9TTM9
Gene Alias [a]1d; ADA1D; Adra1; Adra-1; Adra1a; Adra1d; ADRA1R; Adrd1; adrenergic receptor delta1; adrenergic receptor, alpha 1d; adrenergic, alpha -1D-, receptor; adrenergic, alpha-1A-, receptor; adrenergic, alpha-1D-, receptor; adrenoceptor alpha 1D; alpha 1D-adrenoceptor; alpha 1D-adrenoreceptor; ALPHA1; alpha-1A adrenergic receptor; Alpha-1D adrenergic receptor; alpha-1D adrenoceptor; Alpha-1D adrenoreceptor; alpha-1D-adrenergic receptor; alpha1D-AR; Alpha1delta-AR; alpha-adrenergic receptor 1a; DAR; dJ779E11.2; Gpcr8; RA42; Spr8
Gene Symbols Adra1d
Host Species Rabbit
Immunogen Synthetic 20 amino acid peptide from 3rd cytoplasmic domain of human ADRA1D.
Purification Method Antigen affinity chromatography
Quantity 50 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100009398, 100066003, 146, 539543, 552897, 552898
Target Species Human, Canine, Rabbit, Monkey, Bovine, Pig, Horse, Bat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.