Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alpha 2 antiplasmin Polyclonal Antibody, FITC, Bioss

Rabbit Polyclonal Antibody
Supplier: Bioss BS6188RFITC
This item is not returnable.
View return policy
Description
Alpha 2 antiplasmin Polyclonal antibody specifically detects Alpha 2 antiplasmin in Human, Mouse, Rat samples. It is validated for Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin)Specifications
| Alpha 2 antiplasmin | |
| Polyclonal | |
| FITC | |
| SERPINF2 | |
| A2AP; AAP; AI747498; alpha 2 antiplasmin; alpha-2-antiplasmin; Alpha-2-AP; alpha-2-PI; Alpha-2-plasmin inhibitor; API; pigment epithelium derived factor; plasmin inhibitor alpha 2; Pli; serine (or cysteine) peptidase inhibitor, clade F, member 2; serine (or cysteine) proteinase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2; serine (or cysteine) proteinase inhibitor, clade F, member 2; Serpimf2; serpin F2; serpin family F member 2; serpin peptidase inhibitor, clade F (alpha-2 antiplasmin, pigment epithelium derived factor), member 2; Serpinf2 | |
| KLH conjugated synthetic peptide derived from human SERPINF2/Alpha 2 antiplasmin | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
| Immunocytochemistry, Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin) | |
| 1 mg/mL | |
| TBS with 1% BSA, 50% glycerol and 0.03% ProClin 300 | |
| P08697, Q61247 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 18816, 287527, 5345 | |
| -20° C, store in dark | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction