Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

alpha,2-Dimethylstyrene, 99%

Catalog No. p-7028671
Click to view available options
Quantity:
5 g

CAS: 26444-18-8 | C10H12 | 132.206 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26444-18-8
Molecular Formula C10H12
Molecular Weight (g/mol) 132.206
MDL Number MFCD00049092
InChI Key OGMSGZZPTQNTIK-UHFFFAOYSA-N
Synonym 2-isopropenyltoluene, o-isopropenyltoluene, 1-methyl-2-prop-1-en-2-yl benzene, o,alpha-dimethylstyrene, alpha,2-dimethylstyrene, isopropenyltoluene, o,.alpha.-dimethylstyrene, benzene, 1-methyl-2-1-methylethenyl, o-methyl-.alpha.-methylstyrene, 1-methyl-2-iso-propenylbenzene
PubChem CID 81886
IUPAC Name 1-methyl-2-prop-1-en-2-ylbenzene
SMILES CC1=CC=CC=C1C(=C)C

Specifications

Density 0.894
Boiling Point 53°C to 55°C (11 mmHg)
Flash Point 43°C (110°F)
Refractive Index 1.515
Quantity 5 g
UN Number UN3295
Formula Weight 132.21
Percent Purity 99%
Chemical Name or Material alpha,2-Dimethylstyrene
Hazard Category H226-H315-H319-H335
Hazard Statement GHS H Statement
H226-H315-H319-H335
Flammable liquid and vapour.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P233-P235-P240-P241-P242-P243-P261-P264b-P271-P280-P303+P361+P353-P304+P340-P305+P351+P338-P312-P332+P313-P363-P370+P378q-P501c
DOTInformation Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: HYDROCARBONS, LIQUID, N.O.S.
EINECSNumber 000-000-0
TSCA No
Recommended Storage Keep cold

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.