Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alpha-2-HS-Glycoprotein, Rabbit anti-Human, Polyclonal, Millipore Sigma™

Catalog No. ABN146925UL Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
25 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ABN146925UL 25 μL
ABN1469100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ABN146925UL Supplier MilliporeSigma Supplier No. ABN146925UL
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Anti-Alpha-2-HS-Glycoprotein Antibody, Cat. No. ABN1469, is a highly specific rabbit polyclonal antibody that targets Alpha-2-HS-Glycoprotein and has been tested for use Immunohistochemistry and Western Blotting.

Alpha-2-HS-glycoprotein (UniProt: P02765; also known as Alpha-2-Z-globulin, Ba-alpha-2-glycoprotein, Fetuin-A, Alpha-2 Heremans-Schmid glycoprotein, AHSG) is encoded by the AHSG (also known as FETUA, PRO2743) gene (Gene ID: 197) in human. Alpha-2-HS-glycoprotein is a serum protein of the cystatin superfamily that is synthesized by hepatocytes and is considered as a negative acute-phase reactant. It is highly expressed in various fetal tissues, but in adults it is mainly expressed in the liver. It is synthesized with a signal peptide (aa 1-18) and a connective propeptide (aa 301-340) and is subsequently cleaved into 2 chains known as Alpha-2-HS-glycoprotein chain A and Alpha-2-HS-glycoprotein chain B. The mature product is involved in multiple normal biological and pathological functions. It promotes possesses opsonic properties and is shown to be involved in endocytosis, brain development, bone resorption, regulation of insulin activity and hepatocyte growth factor (HGF) activity. Alpha-2-HS-glycoprotein also serves as a carrier protein and serves as a carrier for otherwise insoluble calcium phosphate and can act as a potent inhibitor of pathological vascular calcification. Higher levels of Alpha-2-HS-glycoprotein have been reported in gestational and type II diabetes. It is also present at higher levels in patients with endometriosis. Lower levels of Alpha-2-HS-glycoprotein are present in cancer subjects. (Ref.: Kalabay, L., et al. (2002). Eur. J. Endocrinol. 147(2); 243-248; Sindhu, S., et al. (2016). BMC Immunol. 17; 33).
TRUSTED_SUSTAINABILITY

Specifications

Antigen Alpha-2-HS-Glycoprotein
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Conjugate Unconjugated
Dilution Western Blotting Analysis: A 1:500 dilution from a representative lot detected Alpha-2-HS-Glycoprotein in human retina tissue lysate.Immunohistochemistry Analysis: A 1:250 dilution from a representative lot detected Alpha-2-HS-Glycoprotein in human liver and human bone marrow tissue sections.
Formulation Purified rabbit polyclonal antibody in buffer containing 0.1 M Tris-Glycine (pH 7.4), 150 mM NaCl with 0.05% sodium azide.
Gene Alias Alpha-2-Z-globulin;Ba-alpha-2-glycoprotein;Fetuin-A;Alpha-2 Heremans-Schmid glycoprotein;AHGS
Gene Symbols AHSG;FETUA;PRO2743
Host Species Rabbit
Immunogen GST/His-tagged recombinant fragment corresponding to 146 amino acids from human Alpha-2-HS-glycoprotein.
Molecular Weight of Antigen ∽55 kDa observed; 39.33 kDa calculated. Uncharacterized bands may be observed in some lysate(s).
Purification Method Affinity Purified
Quantity 25 μL
Regulatory Status RUO
Research Discipline Neuroscience
Primary or Secondary Primary
Gene ID (Entrez) NP_001613.2
Test Specificity This rabbit polyclonal antibody specifically detects human Alpha-2-HS-glycoprotein. It targets an epitope with in the N-terminal half.
Target Species Human
Content And Storage Stable for 1 year at 2-8°C from date of receipt.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.