Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
alpha 2-Macroglobulin Antibody (A2M/3621), DyLight 650, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP308688C
Description
alpha 2-Macroglobulin Monoclonal antibody specifically detects alpha 2-Macroglobulin in Human samples. It is validated for Immunohistochemistry (Paraffin), Protein ArraySpecifications
alpha 2-Macroglobulin | |
Monoclonal | |
DyLight 650 | |
alpha-2-M, alpha-2-macroglobulin, C3 and PZP-like alpha-2-macroglobulin domain-containing protein 5, CPAMD5DKFZp779B086, FWP007, S863-7 | |
Human recombinant alpha 2-Macroglobulin protein fragment (around aa604-748) (exact sequence is proprietary) (Uniprot: P01023) | |
100 μg | |
Neuroscience | |
2 | |
Store at 4°C in the dark. | |
IgG2c κ |
Immunohistochemistry (Paraffin), Protein Array | |
A2M/3621 | |
50mM Sodium Borate | |
Mouse | |
Protein A or G purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction