Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ alpha-2-Macroglobulin, Human Plasma

Catalog No. p-5386179
Click to view available options
Quantity:
1 mg

For research use

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Protease Inhibition
Formulation Lyophilized powder
Molecular Weight (g/mol) 725KDa
Quantity 1 mg
Source Human plasma
Regulatory Status RUO
Product Type alpha-2-Macroglobulin, Human Plasma
Recombinant Native
Content And Storage Ambient temperatures
Form Lyophilized

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.